Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024471.4 | NP_077791.1 | 9 | 1037 | ||
GenBank mRNA | AB057731.1 | - | 9 | 936 | ||
AK031851.1 | - | 9 | 1009 | |||
AK045167.1 | - | 9 | 822 | |||
AK048744.1 | - | 981 | ||||
AK049548.1 | - | 9 | 952 | |||
AK077192.1 | - | 9 | 1038 | |||
BC002141.1 | - | 9 | 969 | 3484559 |