Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086088.1 | - | 5, 6 | 1004 | ||
AK122543.1 | - | 3, 4 | 4208 | |||
AK144928.1 | - | 5, 6 | 1003 | |||
AK150129.1 | - | 5, 6 | 991 | |||
AK156171.1 | - | 5, 6 | 1044 | |||
AK159202.1 | - | 5, 6 | 1034 | |||
AK167203.1 | - | 5, 6 | 1118 | |||
AK172333.1 | - | 5, 6 | 1044 | |||
AK182852.1 | - | 1, 2 | 90 | |||
BC003251.1 | - | 4, 5 | 665 | 3500047 | ||
BC013661.1 | - | 1, 2 | 136 | |||
BC094418.1 | - | 5, 6 | 1026 | 30702568 |