Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286433.1 | NP_001273362.1 | 4 | 557 | ||
NM_001286435.1 | NP_001273364.1 | 5 | 769 | |||
NM_001286436.1 | NP_001273365.1 | 5 | 862 | |||
NM_001286438.1 | NP_001273367.1 | 6 | 837 | |||
NM_001286439.1 | NP_001273368.1 | 6 | 846 | |||
NM_001286440.1 | NP_001273369.1 | 5 | 975 | |||
NM_005009.2 | NP_005000.1 | 5 | 659 | |||
GenBank mRNA | AF086133.1 | - | 3 | 357 | ||
AK094439.1 | - | 2 | 1518 | |||
BC004880.2 | - | 5 | 642 | 3830205 | ||
BC017067.1 | - | 5 | 659 | 3913604 | ||
Y07604.1 | - | 5 | 639 |