Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030556.2 | NP_085033.2 | 4 | 1128 | ||
GenBank mRNA | AF271634.1 | - | 4 | 1128 | ||
AK098112.1 | - | 4 | 1119 | |||
BC109154.1 | - | 4 | 1111 | 40047017 | ||
BC109155.1 | - | 3 | 981 | 40047019 |