Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021079.4 | NP_066565.1 | 6 | 731 | ||
XM_011524858.2 | XP_011523160.1 | 6 | 739 | |||
GenBank mRNA | AF020500.1 | - | 6 | 687 | ||
AF043324.1 | - | 6 | 710 | |||
AK291936.1 | - | 6 | 710 | |||
AK303517.1 | - | 5 | 591 | |||
BC006538.2 | - | 6 | 718 | 3009814 | ||
BC006569.2 | - | 6 | 718 | 3009814 | ||
BC007258.2 | - | 5 | 565 | |||
BC008312.2 | - | 5 | 565 | |||
BC008579.1 | - | 6 | 694 | |||
BU849351.1 | - | 6 | 734 | |||
M86707.1 | - | 5 | 550 |