Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030564.1 | NP_085041.1 | 3 | 450 | ||
GenBank mRNA | AF434815.1 | - | 2 | 345 | ||
AK045696.1 | - | 3 | 446 | |||
AK046274.1 | - | 3 | 432 | |||
AK046381.1 | - | 3 | 446 | |||
AK077221.1 | - | 3 | 447 | |||
AK132330.1 | - | 466 | ||||
AK137606.1 | - | 426 | ||||
AK150976.1 | - | 3 | 444 | |||
AK159753.1 | - | 3 | 432 | |||
BC004042.1 | - | 3 | 450 | 3496314 |