Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159394.1 | NP_001152866.1 | 6, 7 | 1551 | ||
NM_001159395.1 | NP_001152867.1 | 7, 8 | 1433 | |||
NM_030612.3 | NP_085115.1 | 8, 9 | 1781 | |||
GenBank mRNA | AB020974.1 | - | 8, 9 | 1993 | ||
AB026551.2 | - | 6, 7 | 1551 | |||
AB047549.2 | - | 7, 8 | 1446 | |||
AK082908.1 | - | 6, 7 | 1550 | |||
AK136509.1 | - | 3, 4 | 1020 | |||
AK141318.1 | - | 6, 7 | 1552 | |||
AK149946.1 | - | 7, 8 | 1429 | |||
AK150519.1 | - | 6, 7 | 1550 | |||
AK152016.1 | - | 6, 7 | 1552 | |||
AK156826.1 | - | 6, 7 | 1551 | |||
AK159093.1 | - | 6, 7 | 1552 | |||
AK159171.1 | - | 6, 7 | 1552 | |||
AK159459.1 | - | 6, 7 | 1552 | |||
AK159645.1 | - | 6, 7 | 1552 | |||
AK159917.1 | - | 6, 7 | 1552 | |||
AK160065.1 | - | 6, 7 | 1550 | |||
BC058188.1 | - | 6, 7 | 1536 | 5341736 |