Search Thermo Fisher Scientific
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023173.2 | NP_075662.2 | 2 | 534 | ||
GenBank mRNA | AF268196.1 | - | 2 | 459 | ||
AF280810.1 | - | 2 | 401 | |||
AF280811.1 | - | 2 | 401 | |||
AK004488.1 | - | 2 | 416 | |||
AK081862.1 | - | 466 | ||||
AK155026.1 | - | 2 | 1533 | |||
BC099453.1 | - | 2 | 534 | 30938295 |