Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030730.2 | NP_109655.2 | 3 | 563 | ||
GenBank mRNA | AJ132389.1 | - | 3 | 464 | ||
AK083903.1 | - | 3 | 544 | |||
AK143506.1 | - | 2 | 179 | |||
AK155151.1 | - | 3 | 520 | |||
BC133714.1 | - | 3 | 412 | 40143565 | ||
X97574.1 | - | 1 | 67 |