Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000625.4 | NP_000616.3 | 26 | 3599 | ||
XM_011524859.2 | XP_011523161.1 | 26 | 3625 | |||
XM_011524860.2 | XP_011523162.1 | 25 | 3418 | |||
XM_011524861.1 | XP_011523163.1 | 26 | 3525 | |||
XM_011524862.1 | XP_011523164.1 | 24 | 3196 | |||
GenBank mRNA | AB022318.1 | - | 24 | 3218 | ||
AF068236.1 | - | 26 | 3569 | |||
BC130283.1 | - | 26 | 3519 | 40146314 | ||
BC144126.1 | - | 26 | 3519 | 9052646 | ||
D26525.1 | - | 25 | 3349 | |||
L09210.1 | - | 26 | 3541 | |||
L24553.1 | - | 25 | 3411 | |||
U05810.1 | - | 26 | 3529 | |||
U20141.1 | - | 25 | 3335 | |||
U31511.1 | - | 26 | 3440 | |||
X73029.1 | - | 26 | 3560 |