Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CNOT2 SIRNA Assays ›
Gene Symbol:
CNOT2
Gene Name:
CCR4-NOT transcription complex subunit 2
Gene Aliases:
CDC36, HSPC131, NOT2, NOT2H
Chromosome Location:
Chr.12: 70242993 - 70354993 on Build GRCh38
UniGene ID:
Hs.133350
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9625
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CNOT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199302.1
NP_001186231.1
14
1708
NM_001199303.1
NP_001186232.1
13
1961
NM_014515.5
NP_055330.1
13
1839
NR_037615.1
14
2185
XM_006719429.3
XP_006719492.1
14
2064
XM_006719430.3
XP_006719493.1
14
1898
XM_006719432.3
XP_006719495.1
12
1657
XM_006719433.3
XP_006719496.1
10
1429
XM_011538399.1
XP_011536701.1
13
1794
XM_011538400.2
XP_011536702.1
14
2027
XM_017019348.1
XP_016874837.1
15
2085
XM_017019349.1
XP_016874838.1
15
2251
XM_017019350.1
XP_016874839.1
15
2019
XM_017019351.1
XP_016874840.1
15
2185
XM_017019352.1
XP_016874841.1
16
2206
XM_017019353.1
XP_016874842.1
11
1487
XM_017019354.1
XP_016874843.1
11
1653
GenBank mRNA
AF044215.1
-
13
1395
AF113226.1
-
12
1366
AF161480.1
-
13
1377
AF180473.1
-
13
1372
AK000662.1
-
13
1252
AK095789.1
-
13
1924
AK307479.1
-
14
2185
AK315715.1
-
14
1709
AL137674.1
-
5
388
BC002597.2
-
13
1383
3143289
BC011826.2
-
13
1391
3832035
BX641116.1
-
4
3763
Back To Top
More Information
Panther Classification:
Molecular Function -
general transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
nuclear-transcribed mRNA poly(A) tail shortening
trophectodermal cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
positive regulation of cytoplasmic mRNA processing body assembly
negative regulation of translation
gene silencing by RNA
negative regulation of intracellular estrogen receptor signaling pathway
RNA phosphodiester bond hydrolysis, exonucleolytic
regulation of stem cell population maintenance
RNA polymerase II transcription cofactor activity
RNA polymerase II transcription corepressor binding
poly(A)-specific ribonuclease activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-b9vj9:80/10.239.164.80:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0