Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031251.4 | NP_112541.1 | 6 | 489 | ||
GenBank mRNA | AJ250670.1 | - | 5 | 387 | ||
AK054467.1 | - | 7 | 667 | |||
AK078511.1 | - | 6 | 501 | |||
AK079295.1 | - | 6 | 490 | |||
AK085517.1 | - | 7 | 667 | |||
AK137143.1 | - | 6 | 532 | |||
AK170790.1 | - | 6 | 501 | |||
AK170828.1 | - | 6 | 512 | |||
AK189600.1 | - | 1 | 29 | |||
BC005479.1 | - | 6 | 476 | 3484952 | ||
BC006033.1 | - | 6 | 476 |