Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040085.2 | NP_001035174.1 | 13 | 2732 | ||
NM_001040087.2 | NP_001035176.1 | 4 | 939 | |||
NM_001040088.2 | NP_001035177.1 | 4 | 867 | |||
NM_001289583.1 | NP_001276512.1 | 14 | 2780 | |||
NM_001289584.1 | NP_001276513.1 | 14 | 2699 | |||
NM_001289585.1 | NP_001276514.1 | 5 | 1032 | |||
NM_001289586.1 | NP_001276515.1 | 5 | 987 | |||
NM_031394.3 | NP_113571.2 | 13 | 2660 | |||
GenBank mRNA | AB050742.1 | - | 4 | 478 | ||
AB057754.1 | - | 13 | 2119 | |||
AB057755.1 | - | 7 | 2629 | |||
AB057756.1 | - | 5 | 954 | |||
AB057760.1 | - | 13 | 2038 | |||
AB057761.1 | - | 12 | 2071 | |||
AB057762.1 | - | 12 | 1999 | |||
AI746961.1 | - | 4 | 412 | |||
AK027924.1 | - | 4 | 850 | |||
AK051653.1 | - | 5 | 1033 | |||
AK173212.1 | - | 4 | 397 | |||
BC060665.1 | - | 5 | 2482 | |||
BC138714.1 | - | 12 | 1970 | 8861736 | ||
BC145398.1 | - | 13 | 2090 | 9053956 | ||
CA559030.1 | - | 3 | 373 | |||
DV652808.1 | - | 7 | 684 |