Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154337.1 | - | 5, 6 | 1011 | ||
AK009736.1 | - | 5, 6 | 1013 | |||
AK010724.1 | - | 5, 6 | 1025 | |||
AK170103.1 | - | 5, 6 | 1021 | |||
AK171335.1 | - | 5, 6 | 1021 | |||
BC093499.1 | - | 5, 6 | 1008 |
Gene Symbol: Gpr137b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031999.2 | NP_114388.2 | 5, 6 | 1048 | ||
GenBank mRNA | AK050369.1 | - | 5, 6 | 989 | ||
AK080884.1 | - | 5, 6 | 1001 | |||
AK128965.1 | - | 5, 6 | 1048 | |||
AK147921.1 | - | 5, 6 | 1067 | |||
AK172286.1 | - | 5, 6 | 993 | |||
AK185332.1 | - | 1, 2 | 90 | |||
AK193474.1 | - | 1, 2 | 90 | |||
AK196144.1 | - | 1, 2 | 90 | |||
AK199314.1 | - | 1, 2 | 90 | |||
BC003212.1 | - | 5, 6 | 1041 | 3157256 |