Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159986.1 | NP_001153458.1 | 18, 19 | 2300 | ||
NM_033354.3 | NP_203505.3 | 20, 21 | 2547 | |||
GenBank mRNA | AB063356.1 | - | 18, 19 | 2300 | ||
AK132144.1 | - | 18, 19 | 2324 | |||
AK173302.1 | - | 20, 21 | 2528 | |||
BC039223.1 | - | 10, 11 | 1151 | |||
BC051110.1 | - | 11, 12 | 1247 | |||
BC059194.1 | - | 19, 20 | 2365 | 4222915 |