Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053122.4 | NP_444352.2 | 369 | |||
GenBank mRNA | AF359564.1 | - | 301 | |||
AK053361.1 | - | 343 | ||||
AK161351.1 | - | 509 | ||||
BC061049.1 | - | 369 |