Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NPM1 SIRNA Assays ›
Gene Symbol:
NPM1
Gene Name:
nucleophosmin
Gene Aliases:
B23, NPM
Chromosome Location:
Chr.5: 171387648 - 171410900 on Build GRCh38
UniGene ID:
Hs.557550
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9677
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NPM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037738.2
NP_001032827.1
3
443
NM_002520.6
NP_002511.1
3
443
NM_199185.3
NP_954654.1
3
443
XM_005265920.3
XP_005265977.1
3
499
XM_011534564.1
XP_011532866.1
2
263
GenBank mRNA
AB042278.1
-
3
198
AB451236.1
-
3
198
AB451361.1
-
3
198
AK290652.1
-
3
306
AY347529.1
-
4
333
AY740634.1
-
3
270
AY740635.1
-
3
270
AY740636.1
-
3
270
AY740637.1
-
3
270
AY740638.1
-
3
270
AY740639.1
-
3
270
AY740640.1
-
3
270
BC002398.2
-
3
285
2821577
BC008495.1
-
3
295
4276604
BC009623.2
-
3
324
3877633
BC012566.1
-
3
298
4097025
BC014349.1
-
3
294
3511037
BC016716.1
-
3
297
4068167
BC016768.1
-
3
297
4081364
BC016824.1
-
3
297
4106286
BC020467.1
-
3
284
BC021668.1
-
3
298
3930893
BC021983.1
-
3
296
4106023
BC050628.1
-
3
284
5575414
BC107754.1
-
3
299
6736004
BP291033.1
-
3
353
BT007011.1
-
3
198
DQ303464.1
-
3
294
EF429251.1
-
3
198
KU178236.1
-
3
198
KU178237.1
-
3
198
M23613.1
-
3
291
M26697.1
-
3
288
M28699.1
-
3
285
X16934.1
-
3
158
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
nucleosome assembly
intracellular protein transport
nucleocytoplasmic transport
response to stress
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
centrosome cycle
signal transduction
cell aging
protein localization
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of centrosome duplication
viral process
regulation of endodeoxyribonuclease activity
CENP-A containing nucleosome assembly
ribosome assembly
negative regulation of apoptotic process
negative regulation of protein kinase activity by regulation of protein phosphorylation
positive regulation of translation
positive regulation of transcription, DNA-templated
regulation of centriole replication
positive regulation of NF-kappaB transcription factor activity
protein oligomerization
regulation of endoribonuclease activity
regulation of eIF2 alpha phosphorylation by dsRNA
positive regulation of cell cycle G2/M phase transition
transcription coactivator activity
RNA binding
protein kinase inhibitor activity
protein binding
protein kinase binding
Tat protein binding
histone binding
protein homodimerization activity
ribosomal large subunit binding
ribosomal small subunit binding
poly(A) RNA binding
protein heterodimerization activity
NF-kappaB binding
unfolded protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0