Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080560.3 | NP_542127.1 | 2 | 263 | ||
GenBank mRNA | AK005302.1 | - | 2 | 281 | ||
AK005788.1 | - | 2 | 273 | |||
AK161968.1 | - | 2 | 281 | |||
AY039837.1 | - | 2 | 272 | |||
BC034898.1 | - | 2 | 240 | 1382079 | ||
BC067069.1 | - | 2 | 263 | 30550024 | ||
BC132630.1 | - | 2 | 238 | 40130907 | ||
BC138211.1 | - | 2 | 238 | 8861230 | ||
Y09873.1 | - | 2 | 250 |