Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 19 | 3122 | ||
NM_033608.3 | NP_291086.2 | 19 | 3188 | |||
GenBank mRNA | AF317839.1 | - | 19 | 3188 | ||
AK078536.1 | - | 19 | 3117 | |||
AK145121.1 | - | 19 | 3100 | |||
BC034594.1 | - | 18 | 2951 | |||
BC040281.1 | - | 19 | 3148 | 5364073 |