Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 12 | 1751 | ||
NM_033608.3 | NP_291086.2 | 12 | 1817 | |||
GenBank mRNA | AF317839.1 | - | 12 | 1817 | ||
AK078536.1 | - | 12 | 1745 | |||
AK145121.1 | - | 12 | 1726 | |||
BC034594.1 | - | 12 | 1733 | |||
BC040281.1 | - | 12 | 1777 | 5364073 |