Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053136.3 | NP_444366.3 | 1 | 1052 | ||
GenBank mRNA | AK078189.1 | - | 1 | 1053 | ||
AK160803.1 | - | 1 | 1021 | |||
AY013772.1 | - | 1 | 851 |