Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122993.1 | NP_001116465.1 | 4 | 1127 | ||
NM_033149.3 | NP_149161.1 | 3 | 760 | |||
GenBank mRNA | BC047347.1 | - | 4 | 1127 | ||
BC051669.1 | - | 4 | 1127 | |||
BC057887.1 | - | 3 | 760 | 4983830 |