Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079830.2 | NP_001073299.1 | 17, 18 | 3585 | ||
NM_053170.3 | NP_444400.2 | 17, 18 | 3585 | |||
GenBank mRNA | AF220138.1 | - | 2, 3 | 162 | ||
AK129293.1 | - | 17, 18 | 2844 | |||
AY458590.1 | - | 17, 18 | 3591 | |||
BC043471.1 | - | 4, 5 | 580 |