Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053179.3 | NP_444409.1 | 2 | 207 | ||
GenBank mRNA | AB041263.1 | - | 2 | 247 | ||
AK051157.1 | - | 2 | 217 | |||
AK076290.1 | - | 2 | 208 | |||
AK088912.1 | - | 2 | 217 | |||
AK156254.1 | - | 2 | 252 | |||
AK169211.1 | - | 2 | 208 | |||
BC003307.1 | - | 2 | 300 | 3154822 | ||
BC057977.1 | - | 2 | 181 | 5052339 |