Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053199.3 | NP_444429.1 | 5 | 824 | ||
GenBank mRNA | AF195662.1 | - | 5 | 768 | ||
AK036352.1 | - | 6 | 880 | |||
AK038917.1 | - | 5 | 781 | |||
AK053077.1 | - | 5 | 778 | |||
AY059393.1 | - | 5 | 623 | |||
BC029659.1 | - | 2 | 243 | |||
BC079837.1 | - | 6 | 1264 | |||
BC115417.2 | - | 5 | 729 | 40093410 | ||
BC115418.2 | - | 5 | 729 | 40093417 |