Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4833439L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252645.1 | NP_001239574.1 | 8 | 1085 | ||
NM_001252646.1 | NP_001239575.1 | 7 | 978 | |||
NM_001252647.1 | NP_001239576.1 | 6 | 799 | |||
NM_001252648.1 | NP_001239577.1 | 5 | 672 | |||
NM_133797.4 | NP_598558.1 | 9 | 1173 | |||
GenBank mRNA | AK004845.1 | - | 9 | 1132 | ||
AK014792.1 | - | 5 | 606 | |||
AK028697.1 | - | 9 | 1106 | |||
AK145025.1 | - | 9 | 1139 | |||
AK159945.1 | - | 9 | 1136 | |||
AK167616.1 | - | 9 | 1117 | |||
AK169399.1 | - | 9 | 1139 | |||
AK173126.1 | - | 9 | 1086 | |||
AK180100.1 | - | 2 | 166 | |||
AK201111.1 | - | 2 | 166 | |||
AK214948.1 | - | 2 | 166 | |||
AK215307.1 | - | 2 | 166 | |||
AK217329.1 | - | 2 | 166 | |||
BC033445.1 | - | 9 | 1152 | 4222839 | ||
BC039775.1 | - | 9 | 1111 | 4239147 | ||
BC043040.1 | - | 9 | 1129 | 5701249 | ||
BC054417.1 | - | 9 | 1118 | 3969623 |