Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 2 | 332 | ||
GenBank mRNA | AK053450.1 | - | 2 | 359 | ||
AK159374.1 | - | 2 | 388 | |||
AK159552.1 | - | 2 | 382 | |||
AK166576.1 | - | 2 | 381 | |||
AK167604.1 | - | 2 | 323 | |||
AK169250.1 | - | 2 | 307 | |||
AK169390.1 | - | 2 | 331 | |||
AK183165.1 | - | 2 | 118 | |||
BC006877.1 | - | 2 | 332 | 2647513 | ||
BC066172.1 | - | 2 | 326 | 30429192 |