Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133191.2 | NP_573454.2 | 11 | 1047 | ||
GenBank mRNA | BC005492.1 | - | 11 | 1047 | 3487337 | |
BC009098.1 | - | 11 | 1105 | 3482190 |