Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 23 | 2554 | ||
GenBank mRNA | AK036294.1 | - | 23 | 2450 | ||
AK040070.1 | - | 23 | 2509 | |||
AK048552.1 | - | 22 | 2379 | |||
AK054029.1 | - | 23 | 2575 | |||
AK129427.1 | - | 511 | ||||
AK149825.1 | - | 23 | 2381 | |||
BC034516.1 | - | 23 | 2554 | 4458024 | ||
BC047436.1 | - | 11 | 1087 |