Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197082.1 | NP_001184011.1 | 9 | 1024 | ||
NM_133878.3 | NP_598639.1 | 8 | 985 | |||
GenBank mRNA | AK076089.1 | - | 8 | 986 | ||
AK150153.1 | - | 8 | 937 | |||
AK150610.1 | - | 8 | 935 | |||
AK153193.1 | - | 8 | 935 | |||
AK155225.1 | - | 6 | 567 | |||
BC019807.1 | - | 8 | 897 | 4456795 | ||
BC057645.1 | - | 9 | 979 | 3591859 |