Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134791.2 | NP_001128263.1 | 9 | 739 | ||
NM_133885.4 | NP_598646.4 | 12 | 972 | |||
NM_173350.3 | NP_775485.2 | 10 | 778 | |||
GenBank mRNA | AK035658.1 | - | 10 | 727 | ||
AK131984.1 | - | 12 | 943 | |||
AK172010.1 | - | 10 | 765 | |||
AK185731.1 | - | 3 | 277 | |||
AK215553.1 | - | 3 | 279 | |||
BC021507.1 | - | 8 | 600 | 5347121 | ||
BC023759.1 | - | 12 | 927 | |||
BC026927.1 | - | 10 | 661 | 5134780 | ||
BC089163.1 | - | 9 | 680 | 5367603 |