Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242349.1 | NP_001229278.1 | 16 | 1795 | ||
NM_178642.5 | NP_848757.4 | 17 | 1804 | |||
GenBank mRNA | AK029329.1 | - | 3 | 171 | ||
AK032793.1 | - | 12 | 816 | |||
AK052589.1 | - | 17 | 1756 | |||
BC004697.1 | - | 9 | 670 | |||
BC062959.1 | - | 16 | 1681 | 30547439 |