Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam192a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028221.4 | NP_082497.2 | 6 | 703 | ||
GenBank mRNA | AK010053.1 | - | 6 | 668 | ||
AK033988.1 | - | 6 | 634 | |||
AK050704.1 | - | 6 | 638 | |||
AK151603.1 | - | 6 | 657 | |||
AK213161.1 | - | 2 | 56 | |||
BC016088.1 | - | 5 | 443 | 4503452 | ||
BC023158.1 | - | 6 | 542 | 4946201 | ||
BC025472.1 | - | 6 | 679 | 5353148 | ||
BC027060.1 | - | 6 | 665 | 5358022 | ||
BC027818.1 | - | 5 | 471 | 5320967 | ||
BC040818.1 | - | 656 | 4194531 |