Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252479.1 | NP_001239408.1 | 2 | 369 | ||
NM_133978.2 | NP_598739.1 | 2 | 369 | |||
GenBank mRNA | AF188504.1 | - | 2 | 205 | ||
AK040123.1 | - | 2 | 233 | |||
AK151785.1 | - | 2 | 172 | |||
AK167886.1 | - | 2 | 215 | |||
AK168024.1 | - | 2 | 222 | |||
AK168280.1 | - | 2 | 206 | |||
AY241869.1 | - | 2 | 226 | |||
BC038633.1 | - | 2 | 208 | 1066813 |