Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133978.2 | NP_598739.1 | 3 | 555 | ||
GenBank mRNA | AF188504.1 | - | 3 | 391 | ||
AK040123.1 | - | 2 | 1632 | |||
AK151785.1 | - | 3 | 358 | |||
AK167886.1 | - | 3 | 401 | |||
AK168024.1 | - | 3 | 408 | |||
AK168280.1 | - | 3 | 392 | |||
AK215113.1 | - | 1 | 65 | |||
AY241869.1 | - | 3 | 412 |