Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252326.1 | NP_001239255.1 | 8, 9 | 1619 | ||
NM_001252327.1 | NP_001239256.1 | 8, 9 | 1565 | |||
NM_133992.3 | NP_598753.1 | 8, 9 | 1619 | |||
GenBank mRNA | AK004773.2 | - | 8, 9 | 1487 | ||
AK047887.1 | - | 8, 9 | 1579 | |||
AK089001.1 | - | 8, 9 | 1620 | |||
AK129196.1 | - | 8, 9 | 1600 | |||
AK157800.1 | - | 8, 9 | 1523 | |||
BC008521.1 | - | 8, 9 | 1556 | |||
BC075686.1 | - | 8, 9 | 1596 | 30615204 | ||
BC079841.1 | - | 8, 9 | 1580 | 6417806 | ||
CF170307.1 | - | 3, 4 | 476 |