Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013028.2 | NP_001013046.1 | 3 | 586 | ||
GenBank mRNA | AK005698.1 | - | 471 | |||
AK142035.1 | - | 3 | 439 | |||
BC086492.1 | - | 3 | 451 | 30470653 |