Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002764.1 | NP_001002764.1 | 4 | 2178 | ||
GenBank mRNA | AK129203.1 | - | 3 | 797 | ||
BC037179.1 | - | 3 | 530 | |||
BC047279.1 | - | 4 | 2190 | |||
BC066040.1 | - | 4 | 2178 | 6410245 | ||
BC077715.1 | - | 4 | 2141 |