Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134028.2 | NP_598789.1 | 6 | 769 | ||
GenBank mRNA | AB158481.1 | - | 6 | 563 | ||
BC019652.1 | - | 6 | 749 | 4501003 | ||
BC051439.1 | - | 6 | 631 | 6491052 | ||
BC078446.1 | - | 4 | 1204 |