Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134029.2 | NP_598790.1 | 3, 4 | 455 | ||
GenBank mRNA | AK008226.1 | - | 423 | |||
AK035283.1 | - | 454 | ||||
AY061970.1 | - | 3, 4 | 442 | |||
BC020084.1 | - | 3, 4 | 455 | 3986525 |