Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 12 | 945 | ||
NM_001142581.1 | NP_001136053.1 | 11 | 888 | |||
NM_134044.3 | NP_598805.2 | 11 | 902 | |||
GenBank mRNA | AK038165.1 | - | 12 | 913 | ||
AK046322.1 | - | 12 | 916 | |||
AK049643.1 | - | 12 | 892 | |||
AK079055.1 | - | 12 | 892 | |||
AK082261.1 | - | 12 | 873 | |||
AK145043.1 | - | 928 | ||||
BC016646.1 | - | 11 | 811 | 2651233 | ||
BC023716.1 | - | 11 | 913 | 5346489 |