Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146025.1 | NP_001139497.1 | 4 | 776 | ||
NM_001146026.1 | NP_001139498.1 | 4 | 701 | |||
NM_001146027.1 | NP_001139499.1 | 4 | 567 | |||
NM_134064.2 | NP_598825.2 | 4 | 712 | |||
GenBank mRNA | AK147349.1 | - | 4 | 702 | ||
AK160090.1 | - | 4 | 563 | |||
AK160115.1 | - | 4 | 563 | |||
AK161651.1 | - | 4 | 689 | |||
AK171943.1 | - | 4 | 558 | |||
AK172257.1 | - | 4 | 558 | |||
BC017630.1 | - | 4 | 505 | 4507289 | ||
BC035548.1 | - | 4 | 527 | 4221284 | ||
CF728349.1 | - | 4 | 632 | |||
CJ147978.1 | - | 4 | 490 |