Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142152.1 | - | 8 | 999 | ||
AK154645.1 | - | 5 | 3239 | |||
AK163238.1 | - | 6 | 2587 | |||
BC031484.1 | - | 8 | 997 | 2654265 | ||
BC044769.1 | - | 1 | 942 | |||
BC047372.1 | - | 8 | 980 | |||
BC057597.1 | - | 6 | 2565 | |||
BC089379.1 | - | 8 | 954 |