Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024604.2 | NP_001019775.1 | 23 | 2461 | ||
GenBank mRNA | AK133330.1 | - | 16 | 1575 | ||
AK220336.1 | - | 4 | 371 | |||
BC051456.1 | - | 23 | 2455 | 6314094 | ||
BC094609.1 | - | 23 | 2461 | 6834589 |