Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178919.4 | NP_849250.1 | 4 | 668 | ||
GenBank mRNA | AK042427.1 | - | 4 | 628 | ||
AK084606.1 | - | 4 | 494 | |||
AK149916.1 | - | 4 | 620 | |||
AK149934.1 | - | 4 | 669 | |||
AK158371.1 | - | 3 | 721 | |||
AK171488.1 | - | 4 | 649 | |||
BC026448.1 | - | 3 | 448 | |||
BC038039.1 | - | 3 | 1170 | |||
BC058973.1 | - | 2 | 802 | |||
BC079532.1 | - | 4 | 564 | 6403184 |