Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029582.2 | NP_083858.1 | 11, 12 | 2253 | ||
NM_134105.2 | NP_598866.1 | 4, 5 | 496 | |||
GenBank mRNA | AK139620.1 | - | 11, 12 | 2336 | ||
AK155844.1 | - | 11, 12 | 2307 | |||
AK156154.1 | - | 11, 12 | 2294 | |||
AK157288.1 | - | 11, 12 | 2227 | |||
AK193048.1 | - | 1, 2 | 84 | |||
BC019564.1 | - | 4, 5 | 521 | |||
BC020363.1 | - | 10, 11 | 1700 | |||
BC029643.1 | - | 11, 12 | 2253 | 4010810 |