Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000913.5 | NP_000904.1 | 3 | 744 | ||
NM_001200019.1 | NP_001186948.1 | 5 | 852 | |||
NM_001318853.1 | NP_001305782.1 | 4 | 895 | |||
NM_001318854.1 | NP_001305783.1 | 2 | 458 | |||
NM_001318855.1 | NP_001305784.1 | 2 | 501 | |||
NM_182647.3 | NP_872588.1 | 4 | 895 | |||
XM_011528828.2 | XP_011527130.1 | 4 | 1397 | |||
XM_011528830.2 | XP_011527132.1 | 3 | 1132 | |||
XM_017027852.1 | XP_016883341.1 | 3 | 914 | |||
XM_017027853.1 | XP_016883342.1 | 4 | 834 | |||
XM_017027854.1 | XP_016883343.1 | 3 | 769 | |||
XM_017027855.1 | XP_016883344.1 | 4 | 861 | |||
GenBank mRNA | AF115266.1 | - | 2 | 483 | ||
AF126470.1 | - | 2 | 440 | |||
AF348323.1 | - | 3 | 491 | |||
AY268428.1 | - | 2 | 440 | |||
BC038433.1 | - | 5 | 852 | 5164017 | ||
CR542061.1 | - | 2 | 440 | |||
CR542082.1 | - | 2 | 440 | |||
L40949.1 | - | 3 | 691 | |||
U30185.1 | - | 3 | 557 | |||
X77130.1 | - | 3 | 617 |
Set Membership: |
![]() |