Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134126.3 | NP_598887.3 | 6 | 1499 | ||
GenBank mRNA | AK129167.1 | - | 3 | 190 | ||
AK149331.1 | - | 6 | 728 | |||
AK163782.1 | - | 6 | 1127 | |||
BC139005.1 | - | 6 | 728 | 8862027 | ||
BC145494.1 | - | 6 | 728 | |||
DQ266093.1 | - | 6 | 862 |