Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134151.4 | NP_598912.4 | 2 | 326 | ||
GenBank mRNA | AK028785.1 | - | 2 | 188 | ||
AK043837.1 | - | 2 | 250 | |||
AK076634.1 | - | 2 | 171 | |||
AK139579.1 | - | 2 | 171 | |||
AK145356.1 | - | 2 | 219 | |||
AK151880.1 | - | 2 | 178 | |||
AK169708.1 | - | 2 | 102 | |||
BC013552.1 | - | 2 | 257 | 4236514 | ||
BC022143.1 | - | 2 | 221 | 5101324 | ||
BC026615.1 | - | 2 | 233 | 4976999 | ||
BY287648.1 | - | 2 | 326 | |||
BY295788.1 | - | 2 | 326 |