Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025965.3 | NP_080241.3 | 4, 5 | 649 | ||
GenBank mRNA | AF326229.2 | - | 4, 5 | 629 | ||
AF395811.1 | - | 4, 5 | 629 | |||
AK010884.1 | - | 4, 5 | 595 | |||
AK031538.1 | - | 4, 5 | 629 | |||
AK160285.1 | - | 4, 5 | 612 | |||
AK166235.1 | - | 4, 5 | 623 | |||
AK167793.1 | - | 4, 5 | 595 | |||
AK192546.1 | - | 1, 2 | 148 | |||
AY993942.1 | - | 629 | ||||
BC011255.1 | - | 4, 5 | 584 | 4017600 |